Vand, gradient grade for HPLC
Glass bottle, 2,5 L
Supplier: Thermo Fisher Scientific
Vand, gradient grade for HPLC
Catalog # ACRO268300025
Supplier: Thermo Fisher Scientific
CAS Number: 7732-18-5
Jump to:
Product Details & Documents
Synonyms: Hydrogen oxide
- CAS Number:7732-18-5
- Formula:H₂O
- MDL Number:MFCD00011332
- Molecular Weight:18.02 g/mol
- Merck Index:13,10098
- Boiling Pt:100 °C (1013 hPa)
- Melting Pt:0 °C
- Density:1 g/cm³
- Storage Temperature:Stue temperatur
- EINECS:231-791-2
Documents
Specifications
Specification Test Results
- Appearance:Clear colorless Liquid
- Residue after evaporation:≤0.0005 %
- Residue after ignition:≤0.0003 %
- Gradient analysis::
- Absorbance largest eluted peak:≤0.002 A.U.
- Filtered to 0.2 micrometer:Filtered
Specifications
- Pakning:Glass bottle
- Pakn:2,5 L
- Cat. No.:ACRO268300025
- Miljøcertificering:EPA Safer Chemical Solvent (Green Circle Rating); Chem21 Solvent Selection Guide recommended by default and After discussion
- Environmentally Preferable:Sikrere sundhed for mennesker og miljø
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



