Isooctan 99.5% for analysis
Glass bottle, 2,5 L
Leverandør: Thermo Fisher Scientific
Isooctan 99.5% for analysis
Katalog # ACRO265440025
Leverandør: Thermo Fisher Scientific
CAS nummer: 540-84-1
Gå til:
Product Details & Documents
Synonymer: Isobutyltrimethylmethane, Isooctane, Isobutyltrimethylethane
- CAS nummer:540-84-1
- Formel:(CH₃)₂CHCH₂C(CH₃)₃
- MDL nummer:MFCD00008943
- Molecular Weight:114.23 g/mol
- Merck Index:13,05212
- Kogende Pt:98…99 °C (760 mmHg)
- Smeltende Pt:–107 °C
- Tæthed:0,69 g/cm³
- Flash Pt:–12 °C
- Opbevaringstemperatur:Stue temperatur
- EINECS:208-759-1
- FN:1262
- ADR:3,II
Documents
ACRO26544
SDS
Specifikationer
Specifikationstestresultater
- Appearance:Clear liquid
- Infrared spectrum:Conforms
- Water:≤0.01 % (Coulometric)
- GC:≥99.4 %
- Color scale:≤10 APHA
- Residue after evaporation:≤0.001 %
- Aluminium (Al):≤0.5 ppm
- Cadmium (Cd):≤0.1 ppm
- Copper (Cu):≤0.1 ppm
- Iron (Fe):≤0.1 ppm
- Lead (Pb):≤0.1 ppm
- Nickel (Ni):≤0.1 ppm
- Zinc (Zn):≤0.1 ppm
- H2S+foreign org. S cmpds:≤50 ppm
Specifikationer
- Pakning:Glass bottle
- Pakn:2,5 L



