2'-Fluor-4'-methoxyacetophenon 99%
Glass bottle, 5 g
Leverandør: Thermo Fisher Scientific
2'-Fluor-4'-methoxyacetophenon 99%
Katalog # ACRO263540050
Leverandør: Thermo Fisher Scientific
CAS nummer: 74457-86-6
Gå til:
Product Details & Documents
- CAS nummer:74457-86-6
- Formel:C₉H₉FO₂
- MDL nummer:MFCD00042290
- Molecular Weight:168.17
- Kogende Pt:80…85 °C (,1 mmHg)
- Smeltende Pt:50…54 °C
- Flash Pt:>110 °C
- Opbevaringstemperatur:Stue temperatur
Documents
ACRO26354
SDS
Specifikationer
Specifikationstestresultater
- Appearance:White to light yellow Powder or crystals
- Infrared spectrum:Conforms
- Melting point:50 to 54 °C
- GC:≥98.5 %
Specifikationer
- Pakning:Glass bottle
- Pakn:5 g
Specifikationer
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



