Hexacosansyre 98%
Glass bottle for solids, 100 mg
: Cerotic acid, Hexacosanoic acid
- :506-46-7
- :C₂₆H₅₂O₂
- :MFCD00002811
- :396.7 g/mol
- :250 °C
- :87…89 °C
- :0,81 g/cm³
- :>110 °C
- :Køleskab
- :208-040-2
- Appearance:White Glistening powder
- Infrared spectrum:Conforms
- GC:≥97.5 %
- Pakning:Glass bottle for solids
- Pakn:100 mg



