Hexacosansyre 98%
Glass bottle for solids, 1 g
Leverandør: Thermo Fisher Scientific
Gå til:
Product Details & Documents
Synonymer: Cerotic acid, Hexacosanoic acid
- CAS nummer:506-46-7
- Formel:C₂₆H₅₂O₂
- MDL nummer:MFCD00002811
- Molecular Weight:396.7 g/mol
- Kogende Pt:250 °C
- Smeltende Pt:87…89 °C
- Tæthed:0,81 g/cm³
- Flash Pt:>110 °C
- Opbevaringstemperatur:Køleskab
- EINECS:208-040-2
Documents
ACRO25821
SDS
Specifikationer
Specifikationstestresultater
- Appearance:White Glistening powder
- Infrared spectrum:Conforms
- GC:≥97.5 %
Specifikationer
- Pakning:Glass bottle for solids
- Pakn:1 g
Specifikationer
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



