Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Pipette Boxes, with Lid

Pipette Boxes, with Lid

Pipette box, for 23 pipettes of 25 ml, 66×66×460 mm
Katalog # BELC1217-90218
Marketsource
Leverandør:   BELLCO GLASS
Pipette Boxes, with Lid
Katalog # BELC1217-90218
Leverandør:  BELLCO GLASS
Marketsource
Pipette Boxes, with Lid
Pipette Boxes, with Lid

Pipette box, for 23 pipettes of 25 ml, 66×66×460 mm

Price
618,10 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents

Pipette boxes constructed of heavy gauge stainless steel and rectangular. Satin finish on the entire surface for convenient marking and coding.

  • Stackable
  • Silicone pad on inside of top and bottom protect pipette tips
  • Can be sterilized by autoclave or dry heat

Permanently bonded silicone pad on inside of top and bottom protects pipette tips and mouthpieces from chipping.

Specifikationer
  • Beskrivelse:
    Pipette box, for 23 pipettes of 25 ml
  • B×D×H:
    66×66×460 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.