Pipette Boxes, with Lid
Pipette box, for 23 pipettes of 25 ml, 66×66×460 mm
Katalog # BELC1217-22218
Marketsource
Leverandør: BELLCO GLASS
Gå til:
Product Details & Documents
Pipette boxes constructed of heavy gauge stainless steel and rectangular. Satin finish on the entire surface for convenient marking and coding.
- Stackable
- Silicone pad on inside of top and bottom protect pipette tips
- Can be sterilized by autoclave or dry heat
Permanently bonded silicone pad on inside of top and bottom protects pipette tips and mouthpieces from chipping.
Specifikationer
- Beskrivelse:Pipette box, for 23 pipettes of 25 ml
- B×D×H:66×66×460 mm
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



