Pipette Boxes, with Lid
Pipette box, for 81 pipettes of 0,1 - 0,5 ml / 35 pipettes of 10 ml, 66×66×410 mm
Katalog # BELC1217-22216
Marketsource
Leverandør: BELLCO GLASS
Gå til:
Product Details & Documents
Pipette boxes constructed of heavy gauge stainless steel and rectangular. Satin finish on the entire surface for convenient marking and coding.
- Stackable
- Silicone pad on inside of top and bottom protect pipette tips
- Can be sterilized by autoclave or dry heat
Permanently bonded silicone pad on inside of top and bottom protects pipette tips and mouthpieces from chipping.
Specifikationer
- Beskrivelse:Pipette box, for 81 pipettes of 0,1 - 0,5 ml / 35 pipettes of 10 ml
- B×D×H:66×66×410 mm