Orders LinkContactUsLinkComponent
Pipette Boxes, with Lid
Pipette Boxes, with Lid
Pipette box, for 49 pipettes of 0,1 - 2 ml / 9 shortie pipettes of 10 - 12 ml / 49 pasteur pipettes of 9", 50×50×240 mm
Katalog # BELC1217-11109
Marketsource
Leverandør:   BELLCO GLASS
Pipette Boxes, with Lid
Katalog # BELC1217-11109
Leverandør:  BELLCO GLASS
Marketsource
Pipette Boxes, with Lid
Pipette Boxes, with Lid
Pipette box, for 49 pipettes of 0,1 - 2 ml / 9 shortie pipettes of 10 - 12 ml / 49 pasteur pipettes of 9", 50×50×240 mm
Price
490,19 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Pipette boxes constructed of heavy gauge stainless steel and rectangular. Satin finish on the entire surface for convenient marking and coding.
  • Stackable
  • Silicone pad on inside of top and bottom protect pipette tips
  • Can be sterilized by autoclave or dry heat
Permanently bonded silicone pad on inside of top and bottom protects pipette tips and mouthpieces from chipping.
Specifikationer
  • Beskrivelse:
    Pipette box, for 49 pipettes of 0,1 - 2 ml / 9 shortie pipettes of 10 - 12 ml / 49 pasteur pipettes of 9"
  • B×D×H:
    50×50×240 mm