Pipette Boxes, with Lid
Pipette box, for 64 pipettes of 5 ml / 36 shortie pipettes of 10 - 12 ml, 102×102×240 mm
BELC1217-00409
Marketsource
Pipette boxes constructed of heavy gauge stainless steel and rectangular. Satin finish on the entire surface for convenient marking and coding.
- Stackable
- Silicone pad on inside of top and bottom protect pipette tips
- Can be sterilized by autoclave or dry heat
Permanently bonded silicone pad on inside of top and bottom protects pipette tips and mouthpieces from chipping.
- Beskrivelse:Pipette box, for 64 pipettes of 5 ml / 36 shortie pipettes of 10 - 12 ml
- B×D×H:102×102×240 mm