Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

VWR®, Dip Slides

Plate count agar, on both sides, for total count, 10
Catalog # 535122F
Supplier:   VWR Chemicals
VWR®, Dip Slides
Catalog # 535122F
Supplier:  VWR Chemicals
CAS Number:  
VWR®, Dip Slides
VWR®, Dip Slides

Plate count agar, on both sides, for total count, 10

CAS Number:  
Price
148,00 kr
Log in for availability
1 * 10 Items
Product Details & Documents

Dip slides for microbiological monitoring of surfaces and in liquids. Each slide has media on both sides, with a choice of the same medium, or two different media on the two sides of the slide. This allows sampling of two surfaces with every slide. Testing with dip slides is a safe, reliable, economical and fast indicator of the presence of microorganisms. Dip slides, with a push cap, have a larger contact area.

  • Leveres sterile
  • Det tager kun få sekunder at tage prøven
  • Resultaterne kan nemt aflæses efter blot 24-48 timer
Specifications

Specifications

  • Pakn:
    10
  • Beskrivelse:
    Plate count agar, on both sides, for total count
  • Cat. No.:
    535122F
  • Mål:
    30 mm rør, 110 mm høj
  • Kontaktområde:
    10 cm²|10
  • Model:
    Push-on-låg
  • Bakke B×L:
    50×22 mm
  • Bladstørrelse:
    80×28 mm|80×28

Specifications