Chloroform 99+% stabilised for analysis
Glass bottle, 2,5 L
Leverandør: Thermo Fisher Scientific
Chloroform 99+% stabilised for analysis
Katalog # ACRO232090025
Leverandør: Thermo Fisher Scientific
CAS nummer: 67-66-3
Gå til:
Product Details & Documents
Synonymer: Trichlormethan
- CAS nummer:67-66-3
- Formel:CHCl₃
- MDL nummer:MFCD00000826
- Molecular Weight:119.38 g/mol
- Merck Index:13,02160
- Kogende Pt:61 °C (760 mmHg)
- Smeltende Pt:–63 °C
- Tæthed:1,48 g/cm³
- EINECS:200-663-8
- ADR:6.1,III
- :stabiliseret med ethanol
- Opbevaringstemperatur:Stue temperatur
- FN:1888
- NÅ:05-2114614906-45
Documents
Specifikationer
Specifikationstestresultater
- Appearance:Clear liquid
- Infrared spectrum:Conforms
- Water:≤0.01 %
- GC:≥99.0 % (Excluding stabilizer)
- Color scale:≤10 APHA
- Stabilizer:0.6 to 1.0 % ethanol
- Residue after evaporation:≤0.001 %
- Free chlorine:Passes test
- Cadmium (Cd):≤0.1 ppm
- Cobalt (Co):≤0.1 ppm
- Copper (Cu):≤0.1 ppm
- Iron (Fe):≤0.5 ppm
- Nickel (Ni):≤0.1 ppm
- Zinc (Zn):≤0.5 ppm
- Acidity (HCl):≤0.001 %
- Acetone and aldehyde:≤0.005 % (as Acetone)
Specifikationer
- Pakning:Glass bottle
- Pakn:2,5 L



