Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Chloramphenicol 98%

Chloramphenicol 98%

Glass bottle for solids, 25 g
Katalog # ACRO227920250
Sundhed
Leverandør:   Thermo Fisher Scientific
Chloramphenicol 98%
Katalog # ACRO227920250
CAS nummer:  56-75-7
Chloramphenicol 98%
Chloramphenicol 98%

Glass bottle for solids, 25 g

CAS nummer:  56-75-7
Price
482,00 kr
Log ind for tilgængelighed
1 * 25 g
Product Details & Documents
Synonymer:  Chloromycetin®, Tega-Cetin, (R,R)-Chloramphenicol
  • CAS nummer:56-75-7
  • Formel:Cl₂CHCONHCH(CH₂OH)CH(OH)C₆H₄NO₂
  • MDL nummer:MFCD00078159
  • Molecular Weight:323.13 g/mol
  • Merck Index:13,02087
  • Kogende Pt:645 °C (1013 hPa)
  • Smeltende Pt:149…153 °C
  • Tæthed:1,49 g/cm³ (20 °C)
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:200-287-4

Documents

Specifikationer

Specifikationstestresultater

  • Appearance:
    White to yellowish-white to greyish-white Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    149 to 153 °C
  • Loss on drying:
    ≤0.5 % (105 °C)
  • UV spectrophotometry:
    98.0 to 102.0 % (on dried substance)
  • Specific optical rotation:
    +18.5° to +20.5° (20 °C, 589 nm) (c=6, ethanol)

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    25 g

Specifikationer