Saponin
Plastic bottle for solids, 500 g
Leverandør: VWR Chemicals
Gå til:
Product Details & Documents
- CAS nummer:8047-15-2
- MDL nummer:MFCD00081981
- Tæthed:1,015…1.02 g/cm³ (20 °C)
- Opbevaringstemperatur:Stue temperatur
- EINECS:232-462-6
Documents
Certifikater
Søg efter partinummer1L Plastic bottle for solids DIN 60 UN (ZPACK13251000) 20 L Plastic bucket for solids UN Z (ZPACK13247020)
Packaging Information
440914Y_SAPONIN BIOCHEMICAL_BSE_REV0.pdf
BSE/TSE Certificates
Specifikationer
Specifikationstestresultater
- Identification:Passes test
- pH (5 %):4 - 7
- Surface tension (20°C; 0,1%; water):30 - 60 mN/m
Specifikationer
- Pakning:Plastic bottle for solids
- Pakn:500 g
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



