Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Xylitol 99+%

Xylitol 99+%

Plastic bottle, 100 g
Katalog # ACRO225981000
Leverandør:   Thermo Fisher Scientific
Xylitol 99+%
Katalog # ACRO225981000
CAS nummer:  87-99-0
Xylitol 99+%
Xylitol 99+%

Plastic bottle, 100 g

CAS nummer:  87-99-0
Price
659,00 kr
Log ind for tilgængelighed
1 * 100 g
Product Details & Documents
Synonymer:  (2R,3r,4S)-Pentane-1,2,3,4,5-pentaol
  • CAS nummer:87-99-0
  • Formel:C₅H₁₂O₅
  • MDL nummer:MFCD00064292
  • Molecular Weight:152.15 g/mol
  • Merck Index:13,10140
  • Kogende Pt:215…217 °C
  • Smeltende Pt:92…96 °C
  • Tæthed:1,52 g/cm³ (20 °C)
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:201-788-0

Documents

Specifikationer

Specifikationstestresultater

  • Appearance:
    White Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    92 to 96 °C
  • Water:
    ≤0.5 %
  • Sulfated ash:
    ≤0.5 %
  • GC:
    ≤2.0 % Other polyols
  • Arsenic (As):
    ≤3 ppm
  • Heavy metals (as Pb):
    ≤10 ppm
  • Reducing sugars:
    ≤0.2 %

Specifikationer

  • Pakning:
    Plastic bottle
  • Pakn:
    100 g

Specifikationer