Eddikesyre 99.8% for analysis
Glass bottle, 250 mL
Leverandør: Thermo Fisher Scientific
Eddikesyre 99.8% for analysis
Katalog # 222142500.
Leverandør: Thermo Fisher Scientific
CAS nummer: 64-19-7
Gå til:
Product Details & Documents
Synonymer: Vinegar acid, Methanecarboxylic acid, Ethanoic acid
- CAS nummer:64-19-7
- Formel:H₃CCOOH
- MDL nummer:MFCD00036152
- Molecular Weight:60.05 g/mol
- Kogende Pt:117…118 °C (760 mmHg)
- Smeltende Pt:16…16,5 °C
- Tæthed:1,048 g/cm³
- Flash Pt:40 °C
- Opbevaringstemperatur:Stue temperatur
- EINECS:200-580-7
- FN:2789
- ADR:8,II
- NÅ:01-2119475328-30
Documents
Specifikationer
Specifikationstestresultater
- Appearance:Clear liquid
- Water:≤0.2 %
- Color scale:≤10 APHA
- Residue after evaporation:≤0.001 %
- Titration with NaOH:≥99.8 %
- Dilution test:Passes test
- Chloride (Cl):≤1 ppm
- Sulfate (SO4):≤1 ppm
- Arsenic (As):≤0.5 ppm
- Cadmium (Cd):≤0.1 ppm
- Copper (Cu):≤0.1 ppm
- Iron (Fe):≤0.2 ppm
- Nickel (Ni):≤0.1 ppm
- Heavy metals (as Pb):≤0.5 ppm
- Acetic anhydride (C4H6O3):≤0.01 %
- KMnO4-reducing substance:Passes test
- K2Cr2O7-reducing substance:Passes test
Specifikationer
- Pakning:Glass bottle
- Pakn:250 mL



