Eddikesyre 99.8% for analysis
Plastic can, 20 L
: Vinegar acid, Methanecarboxylic acid, Ethanoic acid
- :64-19-7
- :H₃CCOOH
- :MFCD00036152
- :60.05 g/mol
- :117…118 °C (760 mmHg)
- :16…16,5 °C
- :1,048 g/cm³
- :40 °C
- :Stue temperatur
- :200-580-7
- :2789
- :8,II
- :01-2119475328-30
Documents
- Appearance:Clear liquid
- Water:≤0.2 %
- Color scale:≤10 APHA
- Residue after evaporation:≤0.001 %
- Titration with NaOH:≥99.8 %
- Dilution test:Passes test
- Chloride (Cl):≤1 ppm
- Sulfate (SO4):≤1 ppm
- Arsenic (As):≤0.5 ppm
- Cadmium (Cd):≤0.1 ppm
- Copper (Cu):≤0.1 ppm
- Iron (Fe):≤0.2 ppm
- Nickel (Ni):≤0.1 ppm
- Heavy metals (as Pb):≤0.5 ppm
- Acetic anhydride (C4H6O3):≤0.01 %
- KMnO4-reducing substance:Passes test
- K2Cr2O7-reducing substance:Passes test
- Pakning:Plastic can
- Pakn:20 L



