Eddikesyre 99.8% for analysis
Glass bottle, 1 L
: Vinegar acid, Methanecarboxylic acid, Ethanoic acid
- :64-19-7
- :H₃CCOOH
- :MFCD00036152
- :60.05 g/mol
- :117…118 °C (760 mmHg)
- :16…16,5 °C
- :1,048 g/cm³
- :40 °C
- :Stue temperatur
- :200-580-7
- :2789
- :8,II
- :01-2119475328-30
- Appearance:Clear liquid
- Water:≤0.2 %
- Color scale:≤10 APHA
- Residue after evaporation:≤0.001 %
- Titration with NaOH:≥99.8 %
- Dilution test:Passes test
- Chloride (Cl):≤1 ppm
- Sulfate (SO4):≤1 ppm
- Arsenic (As):≤0.5 ppm
- Cadmium (Cd):≤0.1 ppm
- Copper (Cu):≤0.1 ppm
- Iron (Fe):≤0.2 ppm
- Nickel (Ni):≤0.1 ppm
- Heavy metals (as Pb):≤0.5 ppm
- Acetic anhydride (C4H6O3):≤0.01 %
- KMnO4-reducing substance:Passes test
- K2Cr2O7-reducing substance:Passes test
- Pakning:Glass bottle
- Pakn:1 L
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



