Orders LinkContactUsLinkComponent
Inoculating loops and needles, disposable
Inoculating loops and needles, disposable
Platinøse, 1 µl, hvid, 10 loops pr. hylster, 300 hylstre pr. æske
Katalog # 612-9320
Leverandør:   Greiner Bio-One
Inoculating loops and needles, disposable
Katalog # 612-9320
Leverandør:  Greiner Bio-One
Inoculating loops and needles, disposable
Inoculating loops and needles, disposable
Platinøse, 1 µl, hvid, 10 loops pr. hylster, 300 hylstre pr. æske
Price
2.020,00 kr
Log ind for tilgængelighed
1 * 3.000 Items
Product Details & Documents
Flexible polymer.
  • Sterilised by irradiation
  • Individually wrapped
  • Length: 200 mm
Documents
Specifikationer
  • Pakket:
    10 loops pr. hylster, 300 hylstre pr. æske
  • Kapacitet:
    1 µl
  • Farve:
    hvid
  • Beskrivelse:
    Platinøse
  • Seq:
    03