Orders LinkContactUsLinkComponent
Sieves
Stainless steel sieve, 1,00 mm, Sigte MF 1,0
  412-1136
 :   IKA
Sieves
  412-1136
 :  IKA
Sieves
Sieves
Stainless steel sieve, 1,00 mm, Sigte MF 1,0
3.170,00 kr
1 * 1 Items
Interchangeable sieve for insertion into the grinding heads ensures maximum particle size filtering.
  • Model:
    Sigte MF 1,0
  • Beskrivelse:
    Stainless steel sieve, 1,00 mm
  • Seq:
    08