Accessories for Immersion Thermostats EH 4 Basic
Cable PC 2.1, length 5 m, for connecting the thermostat vpc control to a PC (9 pin interface)
Katalog # IKAA2700700
Leverandør: IKA
Gå til:
Product Details & Documents
Specifikationer
- Beskrivelse:Cable PC 2.1, length 5 m, for connecting the thermostat vpc control to a PC (9 pin interface)
- Seq:02