Orders LinkContactUsLinkComponent
Accessories for Immersion Thermostats EH 4 Basic
Accessories for Immersion Thermostats EH 4 Basic
Coated metal hoses, length 1 m, max. teperature 300 °C
Catalog # IKAA2606700
Supplier:   IKA
Accessories for Immersion Thermostats EH 4 Basic
Catalog # IKAA2606700
Supplier:  IKA
Accessories for Immersion Thermostats EH 4 Basic
Coated metal hoses, length 1 m, max. teperature 300 °C
Price
11.430,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
Specifications
  • Beskrivelse:
    Coated metal hoses, length 1 m, max. teperature 300 °C
  • Seq:
    01