Orders LinkContactUsLinkComponent
Pipette tips for labopette®
100 - 1000 µl, Blue, Hirschmann Laborgeräte-, Eppendorf®-, Brand®-, Gilson®-pipetter, Bulkpakket, pose med 1.000 stk.
Catalog # HIRS9490103
Supplier:   HIRSCHMANN
Pipette tips for labopette®
Catalog # HIRS9490103
Supplier:  HIRSCHMANN
Pipette tips for labopette®
Pipette tips for labopette®
100 - 1000 µl, Blue, Hirschmann Laborgeräte-, Eppendorf®-, Brand®-, Gilson®-pipetter, Bulkpakket, pose med 1.000 stk.
Price
198,00 kr
Log in for availability
1 * 1.000 Items
Product Details & Documents
These pipette tips are to be used in conjunction with the labopette® pipette.
  • Free of cadmium
  • Certified conformity
Specifications
  • Pakket:
    Bulkpakket, pose med 1.000 stk.
  • Volumen:
    100 - 1000 µl
  • Seq:
    07
  • Spits farge:
    Blue
  • Til brug med:
    Hirschmann Laborgeräte-, Eppendorf®-, Brand®-, Gilson®-pipetter