Pipette tips for labopette®
100 - 1000 µl, Blue, Hirschmann Laborgeräte-, Eppendorf®-, Brand®-, Gilson®-pipetter, Bulkpakket, pose med 1.000 stk.
Catalog # HIRS9490103
Supplier: HIRSCHMANN
Jump to:
Product Details & Documents
These pipette tips are to be used in conjunction with the labopette® pipette.
- Free of cadmium
- Certified conformity
Specifications
- Pakket:Bulkpakket, pose med 1.000 stk.
- Volumen:100 - 1000 µl
- Seq:07
- Spits farge:Blue
- Til brug med:Hirschmann Laborgeräte-, Eppendorf®-, Brand®-, Gilson®-pipetter