Pipette tips for labopette®
1 - 200 µl, Yellow, Hirschmann Laborgeräte-, Eppendorf®-, Brand®-, Gilson®-pipetter, Bulkpakket, pose med 1.000 stk.
Katalog # HIRS9490102
Leverandør: HIRSCHMANN
Gå til:
Product Details & Documents
These pipette tips are to be used in conjunction with the labopette® pipette.
- Free of cadmium
- Certified conformity
Specifikationer
- Pakket:Bulkpakket, pose med 1.000 stk.
- Volumen:1 - 200 µl
- Seq:02
- Spits farge:Yellow
- Til brug med:Hirschmann Laborgeräte-, Eppendorf®-, Brand®-, Gilson®-pipetter