Orders LinkContactUsLinkComponent
Pasteur pipettes
Pasteur pipettes
Utilproppet, spids ∼120 mm lang, 2,0 ml, 230 mm, 250/hylster, 4 hylstre/æske
Katalog # HIRS9260101
Leverandør:   HIRSCHMANN
Pasteur pipettes
Katalog # HIRS9260101
Leverandør:  HIRSCHMANN
Pasteur pipettes
Pasteur pipettes
Utilproppet, spids ∼120 mm lang, 2,0 ml, 230 mm, 250/hylster, 4 hylstre/æske
Price
582,00 kr
Log ind for tilgængelighed
1 * 1.000 Items
Product Details & Documents
Glass pasteur pipettes with fine drawn tips.
  • With or without cotton plug
  • Short or long form
Certifications: According to ISO 7712.
Specifikationer
  • Pakket:
    250/hylster, 4 hylstre/æske
  • Kapacitet:
    2,0 ml
  • Beskrivelse:
    Utilproppet, spids ∼120 mm lang
  • Længde:
    230 mm