Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Melting point determination tubes

120 mm, 1,35 mm, 0,95 mm
  HIRS9211312
 :   HIRSCHMANN
Melting point determination tubes
  HIRS9211312
Melting point determination tubes
Melting point determination tubes

120 mm, 1,35 mm, 0,95 mm

 
746,00 kr
   
1 * 1.000 Items
 

Klart glas, den ene ende lukket.

 
  • Udv.Ø:
    1,35 mm
  • Ø indv.:
    0,95 mm
  • Længde:
    120 mm
  • Seq:
    28
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.