Stopcocks for burettes, lateral
buretter, 50 ml volumen
Katalog # HIRS8901175
Leverandør: HIRSCHMANN
Gå til:
Product Details & Documents
Lateral burette stopcock, made of borosilicate glass, with glass key, clear glass.
- Excellent chemical resistance
- Excellent temperature resistance
Specifikationer
- Til:buretter, 50 ml volumen
- Seq:03