Propper til standardslib, glas
29/32, Klart glas
Katalog # HIRS8900386
Leverandør: HIRSCHMANN
Gå til:
Product Details & Documents
Stoppers from clear or amber glass.
Certifications: DIN 12252
Specifikationer
- NS:29/32
- Model:Klart glas
Stoppers from clear or amber glass.
Certifications: DIN 12252