Celletællere, Improved Neubauer
Counting chamber with double ruling, bright-lined, with clamps, Imp. Neubauer, 0,1 mm
Katalog # HIRS8110204
Leverandør: HIRSCHMANN
Gå til:
Product Details & Documents
Cell counting chambers designed with double net ruling.
Bestillingsoplysninger: Includes two cover glasses.
Documents
Specifikationer
- Kammerdybde:0,1 mm
- Beskrivelse:Counting chamber with double ruling, bright-lined, with clamps, Imp. Neubauer