Orders LinkContactUsLinkComponent
Celletællere, Improved Neubauer
Celletællere, Improved Neubauer
Counting chamber with double ruling, bright-lined, with clamps, Imp. Neubauer, 0,1 mm
Katalog # HIRS8110204
Leverandør:   HIRSCHMANN
Celletællere, Improved Neubauer
Katalog # HIRS8110204
Leverandør:  HIRSCHMANN
Celletællere, Improved Neubauer
Celletællere, Improved Neubauer
Counting chamber with double ruling, bright-lined, with clamps, Imp. Neubauer, 0,1 mm
Price
1.340,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Cell counting chambers designed with double net ruling.
Bestillingsoplysninger: Includes two cover glasses.
Specifikationer
  • Kammerdybde:
    0,1 mm
  • Beskrivelse:
    Counting chamber with double ruling, bright-lined, with clamps, Imp. Neubauer