Celletællere, Bürker
Counting chamber with clips, bright-lined, Bürker, 0,1 mm
Katalog # 612-5729
Leverandør: HIRSCHMANN
Gå til:
Product Details & Documents
Tællekamre med dobbelt gitterinddeling og inklusiv 2 dækglas til hæmacytometer.
Certifications: In vitro Diagnostic Devices Directive (98/79/EC)
Bestillingsoplysninger: Includes two cover glasses.
Documents
Specifikationer
- Kammerdybde:0,1 mm
- Model:Bürker
- Beskrivelse:Counting chamber with clips, bright-lined