Celletællere, Improved Neubauer
Counting chamber with double ruling, bright-lined, Imp. Neubauer, 0,1 mm
Catalog # HIRS8100204
Supplier: HIRSCHMANN
Jump to:
Product Details & Documents
Cell counting chambers designed with double net ruling.
: Includes two cover glasses.
Specifications
- Kammerdybde:0,1 mm
- Beskrivelse:Counting chamber with double ruling, bright-lined, Imp. Neubauer