Celletællere, Improved Neubauer
Celletæller, Improved Neubauer, dobbelt gitterinddeling, 0,10 mm
Leverandør: HIRSCHMANN
Gå til:
Product Details & Documents
Cell counting chambers designed with double net ruling.
Bestillingsoplysninger: Includes two cover glasses.
Documents
Specifikationer
- Kammerdybde:0,10 mm
- Beskrivelse:Celletæller, Improved Neubauer, dobbelt gitterinddeling