Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Tragte, PP

Tragte, PP

220 mm, 17 mm, 150 mm
Catalog # HIRS7750108
Supplier:   HIRSCHMANN
Tragte, PP
Catalog # HIRS7750108
Supplier:  HIRSCHMANN
Tragte, PP
Tragte, PP

220 mm, 17 mm, 150 mm

Price
466,00 kr
Log in for availability
1 * 12 Items
Product Details & Documents

Funnels, made of PP, translucent. With smooth interior surfaces.

  • Excellent chemical resistance
  • Autoclavable
  • Suitable for use with standard filter paper
Specifications
  • Tragt udv. Ø:
    150 mm
  • Stilk udv. Ø:
    17 mm
  • Stilklængde:
    220 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.