Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Tragte, PP

Tragte, PP

155 mm, 13 mm, 100 mm
Katalog # HIRS7750106
Leverandør:   HIRSCHMANN
Tragte, PP
Katalog # HIRS7750106
Leverandør:  HIRSCHMANN
Tragte, PP
Tragte, PP

155 mm, 13 mm, 100 mm

Price
262,00 kr
Log ind for tilgængelighed
1 * 12 Items
Product Details & Documents

Funnels, made of PP, translucent. With smooth interior surfaces.

  • Excellent chemical resistance
  • Autoclavable
  • Suitable for use with standard filter paper
Specifikationer
  • Tragt udv. Ø:
    100 mm
  • Stilk udv. Ø:
    13 mm
  • Stilklængde:
    155 mm
Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.