Tragte, PP
85 mm, 9 mm, 50 mm
Catalog # HIRS7750104
Supplier: HIRSCHMANN
Jump to:
Product Details & Documents
Funnels, made of PP, translucent. With smooth interior surfaces.
- Excellent chemical resistance
- Autoclavable
- Suitable for use with standard filter paper
Specifications
- Tragt udv. Ø:50 mm
- Stilk udv. Ø:9 mm
- Stilklængde:85 mm
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



