Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Centrifugerør

Centrifugerør

10 ml, ∼16×100 mm, ungraduated
Catalog # HIRS6100101
Supplier:   HIRSCHMANN
Centrifugerør
Catalog # HIRS6100101
Supplier:  HIRSCHMANN
Centrifugerør
Centrifugerør

10 ml, ∼16×100 mm, ungraduated

Price
910,00 kr
Log in for availability
1 * 100 Items
Product Details & Documents

Glass.

  • Graduated or ungraduated
  • Good chemical resistance

Long conical or short conical form.

Specifications
  • Ø×L:
    ∼16×100 mm
  • Model:
    ungraduated
  • Kapacitet:
    10 ml
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.