Automatic burettes, AS, Pellet pattern, lateral stopcocks and intermediate valve
50 ml, 0,1 ml, 500 mm, 0,05 ± ml
HIRS355037560
Automatic burettes, AS, Pellet pattern, lateral stopcocks and intermediate valve
HIRS355037560
Automatic burettes, Pellet pattern, class AS, brown glass, DURAN® with white graduation.
- Main point ring graduation
- Lateral PTFE stopcock with intermediate valve
Certifications: Compliance with DIN EN ISO 385.
- Kapacitet:50 ml
- Inddeling:0,1 ml
- Tolerance:0,05 ± ml
- Scale length:500 mm