Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Burettes, Pellet pattern, brown, lateral stopcock

50 ml, 0,1 ml, 500 mm, 0,075 ± ml
Catalog # HIRS342017560
Supplier:   HIRSCHMANN
Burettes, Pellet pattern, brown, lateral stopcock
Catalog # HIRS342017560
Supplier:  HIRSCHMANN
Burettes, Pellet pattern, brown, lateral stopcock
Burettes, Pellet pattern, brown, lateral stopcock

50 ml, 0,1 ml, 500 mm, 0,075 ± ml

Price
1.890,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents

Automatic burettes, Pellet pattern, class B, DURAN® brown glass, with white graduation.

  • Schellbach stripes for enhanced legibility
  • Lateral stopcock with glass key or PTFE spindle

 : Compliance with DIN EN ISO 385.

Specifications
  • Kapacitet:
    50 ml
  • Inddeling:
    0,1 ml
  • Tolerance:
    0,075 ± ml
  • Scale length:
    500 mm