Burettes, straight stopcock, Schellbach
25 ml, 0,05 ml, 0,045 ± ml
Catalog # 612-3538
Supplier: HIRSCHMANN
Jump to:
Product Details & Documents
DURAN®, klasse B.
- Calibrated to deliver 'Ex'
- Short marks at major graduations, blue graduations
Skalaens længde: 500 mm.
Scale length: 500 mm
Certifications: DIN ISO EN 385
Specifications
- Kapacitet:25 ml
- Inddeling:0,05 ml
- Tolerance:0,045 ± ml