Burettes, straight stopcock, Schellbach
10 ml, 0,02 ml, 0,03 ± ml
Catalog # 612-3537
Supplier: HIRSCHMANN
Jump to:
Product Details & Documents
DURAN®, klasse B.
- Calibrated to deliver 'Ex'
- Short marks at major graduations, blue graduations
Skalaens længde: 500 mm.
Scale length: 500 mm
: DIN ISO EN 385
Specifications
- Kapacitet:10 ml
- Inddeling:0,02 ml
- Tolerance:0,03 ± ml