Burettes, straight stopcock, Schellbach
50 ml, 0,1 ml, 0,075 ± ml
Katalog # 612-3536
Leverandør: HIRSCHMANN
Gå til:
Product Details & Documents
DURAN®, klasse B.
- Calibrated to deliver 'Ex'
- Short marks at major graduations, blue graduations
Skalaens længde: 500 mm.
Scale length: 500 mm
Certifications: DIN ISO EN 385
Specifikationer
- Kapacitet:50 ml
- Inddeling:0,1 ml
- Tolerance:0,075 ± ml