Capillaries for LC systems
Capillaries, non-swaged, green, stainless steel, 500 mm, I.Ø 0,17 mm, to connect pump to autosampler
HEWLG1328-87600
Stainless steel capillaries are made of flexible stainless steel (0,6 mm OD) with ¹/₁₆" OD tubing at both ends to accept standard fittings.
Pre-swaged fittings are assembled according to Swagelok specifications.
- Beskrivelse:Capillaries, non-swaged, green, stainless steel, 500 mm, I.Ø 0,17 mm, to connect pump to autosampler
- Seq:28