Orders LinkContactUsLinkComponent
Supplies for diode array detector (DAD)/ multiple wavelength detector (MWD)
Supplies for diode array detector (DAD)/ multiple wavelength detector (MWD)
Inlet capillary with heat exchanger, 0,12 mm ID, 290 mm long for micro high-pressure flow cell
Katalog # HEWLG1315-87325
Leverandør:   AGILENT HEWLETT PACKARD
Supplies for diode array detector (DAD)/ multiple wavelength detector (MWD)
Katalog # HEWLG1315-87325
Supplies for diode array detector (DAD)/ multiple wavelength detector (MWD)
Supplies for diode array detector (DAD)/ multiple wavelength detector (MWD)
Inlet capillary with heat exchanger, 0,12 mm ID, 290 mm long for micro high-pressure flow cell
Price
1.060,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Specifikationer
  • Beskrivelse:
    Inlet capillary with heat exchanger, 0,12 mm ID, 290 mm long for micro high-pressure flow cell
  • Seq:
    17