Supplies for diode array detector (DAD)/ multiple wavelength detector (MWD)
Inlet capillary with heat exchanger, 0,12 mm ID, 290 mm long for micro high-pressure flow cell
Katalog # HEWLG1315-87325
Leverandør: AGILENT HEWLETT PACKARD
Supplies for diode array detector (DAD)/ multiple wavelength detector (MWD)
Katalog # HEWLG1315-87325
Leverandør: AGILENT HEWLETT PACKARD
Gå til:
Product Details & Documents
Specifikationer
- Beskrivelse:Inlet capillary with heat exchanger, 0,12 mm ID, 290 mm long for micro high-pressure flow cell
- Seq:17