Capillaries for LC systems
Capillaries tubing, stainless steel, 200 mm, 0.12 mm Int.Ø
HEWLG1315-87306
Stainless steel capillaries are made of flexible stainless steel (0,6 mm OD) with ¹/₁₆" OD tubing at both ends to accept standard fittings.
Pre-swaged fittings are assembled according to Swagelok specifications.
- Beskrivelse:Capillaries tubing, stainless steel, 200 mm, 0.12 mm Int.Ø
- Seq:100