Orders LinkContactUsLinkComponent
Capillaries for LC systems
Capillaries for LC systems
Capillaries tubing, stainless steel, 200 mm, 0.12 mm Int.Ø
  HEWLG1315-87306
Capillaries for LC systems
  HEWLG1315-87306
Capillaries for LC systems
Capillaries for LC systems
Capillaries tubing, stainless steel, 200 mm, 0.12 mm Int.Ø
1.060,00 kr
1 * 1 Items
Stainless steel capillaries are made of flexible stainless steel (0,6 mm OD) with ¹/₁₆" OD tubing at both ends to accept standard fittings.
Pre-swaged fittings are assembled according to Swagelok specifications.
  • Beskrivelse:
    Capillaries tubing, stainless steel, 200 mm, 0.12 mm Int.Ø
  • Seq:
    100