Supplies for diode array detector (DAD)/ multiple wavelength detector (MWD)
Outlet capillary, 0,17 mm ID, 200 mm long for standard flow cell, standard flow cell with RFID tag
Catalog # HEWLG1315-87302
Supplier: AGILENT HEWLETT PACKARD
Supplies for diode array detector (DAD)/ multiple wavelength detector (MWD)
Catalog # HEWLG1315-87302
Supplier: AGILENT HEWLETT PACKARD
Jump to:
Product Details & Documents
Specifications
- Beskrivelse:Outlet capillary, 0,17 mm ID, 200 mm long for standard flow cell, standard flow cell with RFID tag
- Seq:15