Orders LinkContactUsLinkComponent
Capillaries for LC systems
Capillaries for LC systems
Capillaries, A/A, stainless steel, to connect damper to purge valve
Katalog # HEWLG1312-67301
Leverandør:   AGILENT HEWLETT PACKARD
Capillaries for LC systems
Katalog # HEWLG1312-67301
Capillaries for LC systems
Capillaries for LC systems
Capillaries, A/A, stainless steel, to connect damper to purge valve
Price
724,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Stainless steel capillaries are made of flexible stainless steel (0,6 mm OD) with ¹/₁₆" OD tubing at both ends to accept standard fittings.
Pre-swaged fittings are assembled according to Swagelok specifications.
Specifikationer
  • Beskrivelse:
    Capillaries, A/A, stainless steel, to connect damper to purge valve
  • Seq:
    17