LC vials and closures
Vials, flat bottom, 6,00 ml, klar, No
Katalog # HEWL9301-1377
Leverandør: AGILENT HEWLETT PACKARD
Gå til:
Product Details & Documents
Specifikationer
- Kasket inkluderet:No
- Kapacitet:6,00 ml
- Farve:klar
- Beskrivelse:Vials, flat bottom
- Seq:02