Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Capillaries for LC systems

Capillary, stainless steel, I.Ø 0,17 mm, 80 mm
Katalog # HEWL79826-87606
Leverandør:   AGILENT HEWLETT PACKARD
Capillaries for LC systems
Katalog # HEWL79826-87606
Capillaries for LC systems
Capillaries for LC systems

Capillary, stainless steel, I.Ø 0,17 mm, 80 mm

Price
kr1,200.00
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents

Stainless steel capillaries are made of flexible stainless steel (0,6 mm OD) with ¹/₁₆" OD tubing at both ends to accept standard fittings.

Pre-swaged fittings are assembled according to Swagelok specifications.

Specifikationer
  • Beskrivelse:
    Capillary, stainless steel, I.Ø 0,17 mm, 80 mm
  • Seq:
    85