Inlet septa
Septa, long-life, non-stick, 11 mm
Catalog # HEWL5183-4761
Supplier: AGILENT HEWLETT PACKARD
Jump to:
Product Details & Documents
These septa are available for a variety of applications and have different upper temperature limits. The lower temperature septa are usually softer, seal better and can withstand more punctures (injections) than their high temperature counterparts.
- Made up low bleed silicone rubber formulations
- Efficient by using sharp syringe needles
- Septa should be replaced to avoid leaks and sample loss
- To be used with recommended temperature range
Specifications
- Ø:11 mm
- Beskrivelse:Septa, long-life, non-stick