Orders LinkContactUsLinkComponent
Inlet septa
Inlet septa
Septa, Advanced Green, non-stick, 5 mm
Katalog # HEWL5183-4760
Leverandør:   AGILENT HEWLETT PACKARD
Inlet septa
Katalog # HEWL5183-4760
Inlet septa
Inlet septa
Septa, Advanced Green, non-stick, 5 mm
Price
829,00 kr
Log ind for tilgængelighed
1 * 50 Items
This product has been blocked by your organization. Please contact your purchasing department for more information
Product Details & Documents
These septa are available for a variety of applications and have different upper temperature limits. The lower temperature septa are usually softer, seal better and can withstand more punctures (injections) than their high temperature counterparts.
  • Made up low bleed silicone rubber formulations
  • Efficient by using sharp syringe needles
  • Septa should be replaced to avoid leaks and sample loss
  • To be used with recommended temperature range
Specifikationer
  • Ø:
    5 mm
  • Beskrivelse:
    Septa, Advanced Green, non-stick